Dowex™ 50WX8, 100-200 mesh, ion-exchange resin
Dowex™ 50WX8, 100-200 mesh, ion-exchange resin
Dowex™ 50WX8, 100-200 mesh, ion-exchange resin
Acros Organics
Dowex™ 50WX8, 100-200 mesh, ion-exchange resin
CAS: 11119-67-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic Bottle
Catalog number ACR335345000
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 50WX8
Physical FormMoist Beads or Spheres
CAS11119-67-8
Particle Size15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
PackagingPlastic Bottle