Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Acros Organics
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR332141000
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydride, 60% dispersion in mineral oil
Melting Point800.0°C
Name NoteIn soluble bags, in resealable cans
CAS7646-69-7
ColorGray to Yellow
View more
Adsorbent, Reduction reagent
RUO – Research Use Only