Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
Methanol-d4, CH4O, CAS Number-811-98-3, methan-d3-ol-d, 2h4 methanol, methyl-d3 alcohol-d, perdeuteromethanol, methanol-d4, tetradeuteromethanol, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d, methyl alcohol-d4, cd3od, 50mL, 01, III, 1187, Colorless, 0.8880g/mL, 99.75% min., 212-378-6 | CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
10 mL
50 mL
Packaging:
Glass Bottle
Catalog number ACR329920250
Price (MYR)
1,027.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,027.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethanol-d4
Density0.8880 g/mL
Deuterium (D)99.75% min.
Flash Point12°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only