Fumonisin B2, 96+%
Fumonisin B2, 96+%
Fumonisin B2, 96+%
Thermo Scientific Chemicals
Fumonisin B2, 96+%
Fumonisin B2, CAS # 116355-84-1, is a diester of tricarballylic acid and is isolated from Fusarium. | CAS: 116355-84-1 | C34H59NO14·3H2O | 759.9 g/mol
Have Questions?
Catalog NumberQuantity
ACR3282600101 mg
Catalog number ACR328260010
Price (MYR)
3,684.00
BOT
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (MYR)
3,684.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFumonisin B2
CAS116355-84-1
Health Hazard 1GHS Signal Word: Danger
Assay Percent Range96+%
Molecular FormulaC34H59NO14·3H2O
View more
General Description
  • Fumonisin B2 is a fumonisin mycotoxin produced by the fungus Fusarium
  • Fumonisin B2 is a structural analog of fumonisin B1 but it is more cytotoxic than B1 and can inhibit sphinganine-N-acetyltransferase (ceramide synthase)
Applications
  • In vitro and in vivo studies have shown Fumonisin B2 can induce apoptosis
  • It can inhibit de novo sphingolipid biosynthesis
  • It is also considered an Aspergillus metabolite
  • It shows cytotoxicity against a panel of seven rat hepatoma cell lines, and MDCK cell line
  • It induces hepatocyte nodule formation, a marker of cancer initiation, in rats
RUO – Research Use Only
General References:
  1. Gelderblom, W.C.A.; Jaskiewicz, K.; Marasas, W.F.O., et al. Fumonisins-novel mycotoxins with cancer-promoting activity produced by Fusarium moniliforme. Appl. Environ. Microbiol. 1988, 54, (7), 1806-1811.
  2. Shier, W.T.; Abbas, H.K.; and Mirocha, C.J. Toxicity of the mycotoxins fumonisins B1 and B2 and Alternaria alternata f. sp. lycopersici toxin (AAL) in cultured mammalian cells. Mycopathologia. 1991, 116, (2), 97-104.
  3. Gelderblom, W.C.A.; Cawood, M.E., Snyman, S.D., et al. Structure-activity relationships of fumonisins in short-term carcinogenesis and cytotoxicity assays. Food Chem. Toxicol. 1993, 31, (6), 407-414.