Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR327081000
100 g
Plastic bottle
Catalog number ACR327081000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium persulfate
Melting Point120.0°C
Name NoteDNAse, RNAse and Protease Free
CAS7727-54-0