Acetone, 99.8%, Extra Dry, AcroSeal™
Acetone, 99.8%, Extra Dry, AcroSeal™
Acetone, 99.8%, Extra Dry, AcroSeal™
Acetone, 99.8%, Extra Dry, AcroSeal™
Acetone, 99.8%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Acetone, 99.8%, Extra Dry, AcroSeal™
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantity
ACR3268000101 L
Catalog number ACR326800010
Price (MYR)
1,024.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,024.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone
Melting Point-95.0°C
Name NoteWater <50 ppm
CAS67-64-1
CAS Min %99.75