Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR326630025
2.5 L
Glass Bottle
Catalog number ACR326630025
Price (MYR)
1,075.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,075.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
Name NoteECD Tested, for Pesticide Analysis
CAS67-56-1
Health Hazard 1Danger