Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
ACR3207700757.5 mL
Catalog number ACR320770075
Price (MYR)
524.00
BOT
Add to cart
Quantity:
7.5 mL
Request bulk or custom format
Price (MYR)
524.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Density1.1900g/mL
Deuterium (D)99.85% min.
Flash Point88°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only