Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific™
Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific™
Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific™
Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific™
Thermo Scientific Chemicals
Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific™
Have Questions?
Catalog number ACR319270010
Price (MYR)
592.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
592.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl 3-isocyanatopropionate
CAS5100-34-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.