Thulium(III) chloride, 99.9%, (trace metal basis)
Thulium(III) chloride, 99.9%, (trace metal basis)
Thulium(III) chloride, 99.9%, (trace metal basis)
Thulium(III) chloride, 99.9%, (trace metal basis)
Acros Organics
Thulium(III) chloride, 99.9%, (trace metal basis)
CAS: 13537-18-3 | Cl3Tm | 275.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3160300101 gGlass bottle
Catalog number ACR316030010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThulium(III) chloride
Melting Point821°C
Name Note99.9%
CAS13537-18-3
ColorWhite
View more
for ultra trace analysis
RUO – Research Use Only