Oxytetracycline hydrochloride, can be used as secondary standard'
Oxytetracycline hydrochloride, can be used as secondary standard'
Oxytetracycline hydrochloride, can be used as secondary standard'
Oxytetracycline hydrochloride, can be used as secondary standard'
Thermo Scientific Chemicals
Oxytetracycline hydrochloride, can be used as secondary standard'
Have Questions?
Catalog NumberQuantity
ACR301275000500 mg
Catalog number ACR301275000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialOxytetracycline hydrochloride
TypeOxytetracycline Hydrochloride
Physical FormCrystalline Powder
CAS2058-46-0
Health Hazard 1LIGHT SENSITIVE