BOC-L-Proline, 99+%
BOC-L-Proline, 99+%
BOC-L-Proline, 99+%
Thermo Scientific Chemicals
BOC-L-Proline, 99+%
CAS: 15761-39-4 | C10H17NO4 | 215.249 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR27565025025 gGlass Bottle
Catalog number ACR275650250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBOC-L-Proline
Melting Point133.0°C to 136.0°C
CAS15761-39-4
ColorWhite
Infrared SpectrumAuthentic