BOC-L-Alanine, 99+%
BOC-L-Alanine, 99+%
BOC-L-Alanine, 99+%
Thermo Scientific Chemicals
BOC-L-Alanine, 99+%
CAS: 15761-38-3 | C8H15NO4 | 189.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR27549050050 gGlass Bottle
Catalog number ACR275490500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBOC-L-Alanine
Melting Point81.0°C to 84.0°C
CAS15761-38-3
ColorWhite
Infrared SpectrumAuthentic