Sodium oxalate, 99.5%, for analysis, Thermo Scientific Chemicals
Sodium oxalate, 99.5%, for analysis, Thermo Scientific Chemicals
Sodium oxalate, 99.5%, for analysis, Thermo Scientific Chemicals
Sodium oxalate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium oxalate, 99.5%, for analysis, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR2705400101 kg
Catalog number ACR270540010
Price (MYR)
1,188.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,188.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium oxalate
Name Notep.a.
CAS62-76-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: