Toluene, 99.85%, for HPLC
Toluene, 99.85%, for HPLC
Toluene, 99.85%, for HPLC
Toluene, 99.85%, for HPLC
Thermo Scientific Chemicals
Toluene, 99.85%, for HPLC
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2683700252.5 LGlass Bottle
Catalog number ACR268370025
Price (MYR)
611.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
611.00
EA
Add to cart
Specifications
Chemical Name or MaterialToluene
Absorbance(1 cm cell vs HPLC−grade water), 0.01 max. at 350nm, 0.02 max. at 340nm, 0.03 max. at 330nm, 0.05 max. at 310nm, 0.05 max. at 320nm, 0.1 max. at 300nm, 0.5 max. at 290nm, cut off at 286nm
CAS108-88-3
Health Hazard 1Signal Word: DANGER!
Acidity0.0015 meq/g max.
View more
For HPLC
RUO – Research Use Only