Lead(II) perchlorate trihydrate, 99%
Lead(II) perchlorate trihydrate, 99%
Lead(II) perchlorate trihydrate, 99%
Lead(II) perchlorate trihydrate, 99%
Thermo Scientific Chemicals
Lead(II) perchlorate trihydrate, 99%
CAS: 13453-62-8 | Cl2H6O11Pb | 460.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR254592500250 gGlass bottle
Catalog number ACR254592500
Price (MYR)
1,321.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,321.00
EA
Add to cart
Specifications
Chemical Name or MaterialLead(II) perchlorate trihydrate
Melting Point100.0°C
Name Note99%
CAS13453-62-8
ColorWhite to Beige
View more
Oxidation reagent
RUO – Research Use Only