Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Thermo Scientific Chemicals
Theophylline, 99+%, anhydrous
CAS: 58-55-9 | C7H8N4O2 | 180.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR250312500
Price (MYR)
779.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
779.00
EA
Add to cart
Specifications
Chemical Name or MaterialTheophylline
Name Note99+%
CAS58-55-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: