Chromium, 99+%, pure, powder
Chromium, 99+%, pure, powder
Chromium, 99+%, pure, powder
Chromium, 99+%, pure, powder
Thermo Scientific Chemicals
Chromium, 99+%, pure, powder
CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Catalog NumberQuantity
ACR225491000100 g
Catalog number ACR225491000
Price (MYR)
691.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
691.00
EA
Add to cart
Specifications
Chemical Name or MaterialChromium
CAS7440-47-3
Melting Point1857.0°C
Boiling Point2642.0°C
Health Hazard 1Warning