Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Thermo Scientific Chemicals
Lead(II,IV) oxide, red, 98%
CAS: 1314-41-6 | O4Pb3 | 685.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR221115000500 gGlass bottle
Catalog number ACR221115000
Price (MYR)
458.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
458.00
EA
Add to cart
Specifications
Chemical Name or MaterialLead(II, IV) oxide, red
Melting Point500.0°C
Name Note98%
CAS1314-41-6
ColorOrange
View more
Suitable for battery materials development.
RUO – Research Use Only