View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydroquinone bis(2-hydroxyethyl)ether
Name Note99+%
CAS104-38-1
Health Hazard 1HYGROSCOPIC
Melting Point104.0°C to 107.0°C