Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
Thermo Scientific Chemicals
Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
CAS: 666-52-4 | C3H6O | 64.117 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR217371000100 mLAcroSeal™ Glass Bottle
Catalog number ACR217371000
Price (MYR)
3,201.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
3,201.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone-d6
Density0.8700g/mL
Deuterium (D)99.75% min.
Flash Point−20°C
PackagingAcroSeal™ Glass Bottle
View more
For NMR
RUO – Research Use Only