Phosphorus pentoxide, 99+%, for analysis
Phosphorus pentoxide, 99+%, for analysis
Phosphorus pentoxide, 99+%, for analysis
Phosphorus pentoxide, 99+%, for analysis
Thermo Scientific Chemicals
Phosphorus pentoxide, 99+%, for analysis
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR215755000
Price (MYR)
808.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
808.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhosphorus pentoxide
Melting Point340.0°C to 360.0°C
Name Notep.a.
CAS1314-56-3
ColorWhite
View more
Adsorbent
RUO – Research Use Only