Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Thermo Scientific Chemicals
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
CAS: 21351-79-1 | CsHO | 149.91 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR213601000100 gPlastic bottle
Catalog number ACR213601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCesium hydroxide
Name Note99%
CAS21351-79-1
ColorColorless
Health Hazard 1GHS Signal Word: Danger
View more
For ultra trace analysis
RUO – Research Use Only