Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Thermo Scientific Chemicals
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
CAS: 12092-47-6 | C16H24Cl2Rh2 | 493.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR211305000500 mgGlass bottle
Catalog number ACR211305000
Price (MYR)
4,679.00
EA
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,679.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloro(1, 5-cyclooctadiene)rhodium(I) dimer
Name Notemin. 40.8% Rh
CAS12092-47-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement: