4-Aminobenzotrifluoride, 99%
4-Aminobenzotrifluoride, 99%
4-Aminobenzotrifluoride, 99%
4-Aminobenzotrifluoride, 99%
Thermo Scientific Chemicals
4-Aminobenzotrifluoride, 99%
CAS: 455-14-1 | C7H6F3N | 161.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR20891010010 mLGlass bottle
Catalog number ACR208910100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminobenzotrifluoride
Name Note99%
CAS455-14-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger