Amberlite™ IR-120 Na ion-exchange resin
Amberlite™ IR-120 Na ion-exchange resin
Amberlite™ IR-120 Na ion-exchange resin
Amberlite™ IR-120 Na ion-exchange resin
Amberlite™ IR-120 Na ion-exchange resin
CAS: 9002-23-7 | (C10H11)A(C8H9O3S)BC2H6
Have Questions?
Catalog NumberQuantityPackaging
ACR208412500250 gPlastic bottle
Catalog number ACR208412500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IR120
Physical FormBeads
CAS9002-23-7
Particle SizeParticle size analysis: uniformity coefficient: <1.9, harmonic mean size: 0.600 to 0.800mm,% <0.300mm: <2.0%
PackagingPlastic bottle
View more
Ion exchange resin
RUO – Research Use Only