Ammonium thiocyanate, 99+%, extra pure
Ammonium thiocyanate, 99+%, extra pure
Ammonium thiocyanate, 99+%, extra pure
Ammonium thiocyanate, 99+%, extra pure
Thermo Scientific Chemicals
Ammonium thiocyanate, 99+%, extra pure
CAS: 1762-95-4 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR206505000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium thiocyanate
Melting Point149.0°C
CAS1762-95-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger