Aluminum ammonium sulfate dodecahydrate, 99%, pure
Aluminum ammonium sulfate dodecahydrate, 99%, pure
Aluminum ammonium sulfate dodecahydrate, 99%, pure
Aluminum ammonium sulfate dodecahydrate, 99%, pure
Acros Organics
Aluminum ammonium sulfate dodecahydrate, 99%, pure
CAS: 7784-26-1 | H4AlNO8S2·12H2O | 453.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number ACR206260100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Boiling Point200.0°C
Chemical Name or MaterialAluminum ammonium sulfate dodecahydrate
Melting Point93.5°C
CAS7784-26-1