Aluminum acetylacetonate, 97%
Aluminum acetylacetonate, 97%
Aluminum acetylacetonate, 97%
Aluminum acetylacetonate, 97%
Thermo Scientific Chemicals
Aluminum acetylacetonate, 97%
CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR196975000500 gPlastic bottle
Catalog number ACR196975000
Price (MYR)
1,222.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,222.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum acetylacetonate
CAS13963-57-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0