Iodine monochloride, 98%
Iodine monochloride, 98%
Iodine monochloride, 98%
Iodine monochloride, 98%
Thermo Scientific Chemicals
Iodine monochloride, 98%
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR196965000
Price (MYR)
2,357.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,357.00
EA
Add to cart
Specifications
Boiling Point97.0°C
Chemical Name or MaterialIodine monochloride
Melting Point26.0°C
Name Note98%
CAS7790-99-0
View more
C-X bond formation reagent
RUO – Research Use Only