Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Acros Organics
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
CAS: 8017-16-1 | (HO3P)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number ACR196950100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyphosphoric acid
CAS8017-16-1
Boiling Point550.0°C
Density2.0500g/mL
Recommended StorageMay darken over time