Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Thermo Scientific Chemicals
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
ACR196465000500 mL
Catalog number ACR196465000
Price (MYR)
528.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
528.00
BOT
Add to cart
Chemical Identifiers
CAS7440-66-6, 7647-01-0, 7732-18-5, 7646-85-7
Specifications
Chemical Name or MaterialZinc standard solution
Name Note1 mg/ml Zn in 2% HCl
PackagingPlastic bottle
CAS7440-66-6
Health Hazard 1GHS Signal Word: Warning
View more
for atomic adsorption spectroscopy
RUO – Research Use Only