Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Thermo Scientific Chemicals
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Catalog NumberQuantity
ACR195805000500 mL
Catalog number ACR195805000
Price (MYR)
581.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
581.00
BOT
Add to cart
Chemical Identifiers
CAS7784-27-2, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialAluminum standard solution
Name Note1 mg/ml Al in 0.5N HNO3
PackagingPlastic bottle
CAS7784-27-2
Health Hazard 1Danger
View more
For atomic adsorption spectroscopy
RUO – Research Use Only