Poly(ethylene glycol methyl ether), average M.W. 750
Poly(ethylene glycol methyl ether), average M.W. 750
Poly(ethylene glycol methyl ether), average M.W. 750
Poly(ethylene glycol methyl ether), average M.W. 750
Acros Organics
Poly(ethylene glycol methyl ether), average M.W. 750
CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
Packaging:
Glass bottle
Catalog number ACR192320050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(ethylene glycol methyleether)
ColorWhite
Name NoteAverage M.W. 750
pH4 to 7 (5% aq. soln.)
CAS9004-74-4