2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals
2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals
2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals
2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals
2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR18909050050 gPlastic bottle
Catalog number ACR189090500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-(4-Chlorophenoxy)-2-methylpropionic acid
Name Note99%
CAS882-09-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning