Poly(ethylene oxide), M.W. 200,000 to 300,000
Poly(ethylene oxide), M.W. 200,000 to 300,000
Poly(ethylene oxide), M.W. 200,000 to 300,000
Poly(ethylene oxide), M.W. 200,000 to 300,000
Thermo Scientific Chemicals
Poly(ethylene oxide), M.W. 200,000 to 300,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number ACR178592500
Price (MYR)
2,789.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,789.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(ethylene oxide)
ColorWhite
Name NoteApproximate M.W. 200.000
CAS25322-68-3
Physical FormGranules or Crystalline Powder