2-chloro-5-methylphenol, 99%
2-chloro-5-methylphenol, 99%
2-chloro-5-methylphenol, 99%
2-chloro-5-methylphenol, 99%
Acros Organics
2-chloro-5-methylphenol, 99%
CAS: 615-74-7 | C7H7ClO | 142.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR170411000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-chloro-5-methylphenol
CAS615-74-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0