D(+)-Tryptophan, 99%
D(+)-Tryptophan, 99%
D(+)-Tryptophan, 99%
D(+)-Tryptophan, 99%
Acros Organics
D(+)-Tryptophan, 99%
CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number ACR168635000
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Tryptophan
Melting Point282.0°C to 285.0°C
CAS153-94-6
ColorWhite to Yellow
Infrared SpectrumAuthentic