1-Octanethiol, 97%
1-Octanethiol, 97%
1-Octanethiol, 97%
Thermo Scientific Chemicals
1-Octanethiol, 97%
CAS: 111-88-6 | C8H18S | 146.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR157482500
Price (MYR)
470.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
470.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octanethiol
CAS111-88-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0