Ethyl salicylate, 99+%, Thermo Scientific Chemicals
Ethyl salicylate, 99+%, Thermo Scientific Chemicals
Ethyl salicylate, 99+%, Thermo Scientific Chemicals
Ethyl salicylate, 99+%, Thermo Scientific Chemicals
Ethyl salicylate, 99+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR1504900505 gGlass bottle
Catalog number ACR150490050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl salicylate
Name Note99+%
Acidity1mg KOH/g max.
CAS118-61-6
Health Hazard 1GHS Signal Word: Warning