2-Methyl-2-propanethiol, 99%
2-Methyl-2-propanethiol, 99%
2-Methyl-2-propanethiol, 99%
2-Methyl-2-propanethiol, 99%
Acros Organics
2-Methyl-2-propanethiol, 99%
CAS: 75-66-1 | C4H10S | 90.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR149025000
Price (MYR)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-2-propanethiol
Name Note99%
CAS75-66-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger