Butyric acid hydrazide, 95%, Thermo Scientific™
Butyric acid hydrazide, 95%, Thermo Scientific™
Butyric acid hydrazide, 95%, Thermo Scientific™
Butyric acid hydrazide, 95%, Thermo Scientific™
Butyric acid hydrazide, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ACR14746025025 g
Catalog number ACR147460250
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialButyric acid hydrazide
CAS3538-65-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a