1,4-Pentadiene, 99%
1,4-Pentadiene, 99%
1,4-Pentadiene, 99%
Thermo Scientific Chemicals
1,4-Pentadiene, 99%
CAS: 591-93-5 | C5H8 | 68.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1298600505 gGlass bottle
Catalog number ACR129860050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Pentadiene
Name Note99%
CAS591-93-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger