1-Naphthyl isothiocyanate, 98%
1-Naphthyl isothiocyanate, 98%
1-Naphthyl isothiocyanate, 98%
1-Naphthyl isothiocyanate, 98%
Thermo Scientific Chemicals
1-Naphthyl isothiocyanate, 98%
CAS: 551-06-4 | C11H7NS | 185.25 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR12824010010 gGlass bottle
Catalog number ACR128240100
Price (MYR)
1,041.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,041.00
EA
Add to cart
Specifications
Chemical Name or Material1-Naphthyl isothiocyanate
Name Note98%
CAS551-06-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger