1,3-Cyclohexadiene, 96%, stabilized
1,3-Cyclohexadiene, 96%, stabilized
1,3-Cyclohexadiene, 96%, stabilized
1,3-Cyclohexadiene, 96%, stabilized
Thermo Scientific Chemicals
1,3-Cyclohexadiene, 96%, stabilized
CAS: 592-57-4 | C6H8 | 80.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR11110025025 mLGlass bottle
Catalog number ACR111100250
Price (MYR)
1,714.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,714.00
EA
Add to cart
Specifications
Chemical Name or Material1,3-Cyclohexadiene
Name Note96%
CAS592-57-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger