Acethydrazide, 95%
Acethydrazide, 95%
Acethydrazide, 95%
Thermo Scientific Chemicals
Acethydrazide, 95%
CAS: 1068-57-1 | C2H6N2O | 74.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR10235025025 gGlass bottle
Catalog number ACR102350250
Price (MYR)
529.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
529.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcethydrazide
Name Note95%
CAS1068-57-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning