A2 and A5 Large Gel Electrophoresis System Standard Comb
A2 and A5 Large Gel Electrophoresis System Standard Comb
Actual product may vary
A2 and A5 Large Gel Electrophoresis System Standard Comb
A2 and A5 Large Gel Electrophoresis System Standard Comb
A2 and A5 Large Gel Electrophoresis System Standard Comb
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs, for use with Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
A2-MT2DMicrowell Comb42 (2X)1.5 mm
A2-20DStandard Comb201.5 mm
A2-28CStandard Comb281 mm
Catalog number A2-MT2D
Price (MYR)
510.00
EA
Add to cart
Type:
Microwell Comb
Teeth Count:
42 (2X)
Thickness (Metric) Teeth:
1.5 mm
Price (MYR)
510.00
EA
Add to cart
Eliminate comb assembly with an assortment of heavy-duty, one-piece Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems Combs, for use with Owl A2 and A5 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
For Use With (Equipment)Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems
TypeMicrowell Comb
Teeth Count42 (2X)
Thickness (Metric) Teeth1.5 mm
Unit SizeEA