Owl™ A2 Large Gel Systems
Owl™ A2 Large Gel Systems
Actual product may vary
Owl™ A2 Large Gel Systems
Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Have Questions?
Catalog NumberType
A2-BPLarge, w/Buffer Exchange Ports
Catalog number A2-BP
Price (MYR)
6,578.00
EA
Add to cart
Type:
Large, w/Buffer Exchange Ports
Price (MYR)
6,578.00
EA
Add to cart
Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Features:
  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Includes:
  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (16-, 24- and 36-well), 1.5mm thick
Specifications
Certifications/ComplianceCE marked
DescriptionComplete system with buffer exchange ports
Product LineOwl
TypeLarge, w/Buffer Exchange Ports
Sample Size8 to 432 (16-, 24- and 36-well combs)
Volume (Metric) Buffer2.3 L
Dimensions (L x W x H)14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
Length (English)14.8 in.
Length (Metric)37.5 cm
Length (Metric) Gel25 cm
Width (English)12.8 in.
Width (Metric)32.5 cm
Width (Metric) Gel20 cm
Height (English)4.1 in.
Height (Metric)10.5 cm
Unit SizeEA
Frequently asked questions (FAQs)
Is the PowerEase Touch Power Supply (Cat. No. PS0351) compatible with the Owl gel Systems?
Yes, it is compatible with the Owl Gel Systems. The following ones can be used with the PowerEase power supplies with no need for adaptors:
    Owl EasyCast B1 Mini Gel Electrophor. Sys. (Cat. No. B1-BP)
    Owl A2 Large Gel Sys. (Cat. No. A2)
    Owl EasyCast B3 Gel Sys. with Built-In Recirculat. (Cat. No. B3).
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.