Check Order Status
Bottom-of-the-Bottle™ Filter/Sparger
Bottom-of-the-Bottle™ Filter/Sparger
Small enough to fit narrow neck bottles
Have Questions?
Catalog number
A-437
Price (MYR)
-
Bottom-of-the-Bottle is a trademark of IDEX Health & Science, LLC
Specifications
For Use With (Equipment)
Configured for 1/8 in. O.D. Tubing
Material
PEEK
Pore Size
2 μm
Type
Filter/Sparger
Diameter (Metric) Inner
23 mm