Bath Circulator Fluids
Product Image
Bath Circulator Fluids
Use this transparent bath circulator fluid with our refrigerated circulating baths and chillers.
Have Questions?
Catalog NumberCapacity (Metric)ViscosityDescriptionTemperature Range
999-020410 LDynamic - 11 cP at 20 °C, Kinematic - 11.8 cSt at 20 °CSpecific heat - 0.36 Cal/g °C at 20 °C, Density - 0.93 g/cm3 at 20 °C-40°C to +200°C
Catalog number 999-0204
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
10 L
Viscosity:
Dynamic - 11 cP at 20 °C, Kinematic - 11.8 cSt at 20 °C
Description:
Specific heat - 0.36 Cal/g °C at 20 °C, Density - 0.93 g/cm3 at 20 °C
Temperature Range:
-40°C to +200°C
Use this transparent bath circulator fluid with Thermo Scientific™ VersaCool™ Refrigerated Circulating Baths and Recirculating Chillers in a variety of temperature ranges.
Color: Transparent
Specifications
ColorTransparent
DescriptionSpecific heat - 0.36 Cal/g °C at 20 °C, Density - 0.93 g/cm3 at 20 °C
Capacity (Metric)10 L
Temperature Range-40°C to +200°C
TypeSilicone Oil
ModelSil 180
Quantity10 L
ViscosityDynamic - 11 cP at 20 °C, Kinematic - 11.8 cSt at 20 °C
Unit SizeEA